Assay Details
| Gene Symbol: GUK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000858.5 | NP_000849.1 | 2 - 3 | 307 | 107 | |
| XM_005273105.3 | XP_005273162.1 | 2 - 3 | 335 | 107 | ||
| XM_017001093.1 | XP_016856582.1 | 2 - 3 | 274 | 107 | ||
| XM_017001095.1 | XP_016856584.1 | 2 - 3 | 271 | 107 | ||
| GenBank mRNA | BC006249.2 | - | 2 - 3 | 245 | 3946293 | 107 |
| BC009914.2 | - | 2 - 3 | 219 | 2821563 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC006249.2, BC009914.2, NM_000858.5, XM_005273105.3, XM_017001093.1, XM_017001095.1 transcripts.
Set Membership: |
|

GUK1
Genomic Map