Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011544021.2 | XP_011542323.1 | 6 - 7 | 724 | 86 | |
XM_011544022.2 | XP_011542324.1 | 6 - 7 | 799 | 86 | ||
XM_011544023.2 | XP_011542325.1 | 6 - 7 | 724 | 86 | ||
XM_011544024.2 | XP_011542326.1 | 6 - 7 | 724 | 86 | ||
XM_011544026.2 | XP_011542328.1 | 6 - 7 | 724 | 86 | ||
XM_011544029.2 | XP_011542331.1 | 6 - 7 | 724 | 86 | ||
XM_017000103.1 | XP_016855592.1 | 6 - 7 | 724 | 86 | ||
XM_017000108.1 | XP_016855597.1 | 6 - 7 | 799 | 86 | ||
GenBank mRNA | AK023586.1 | - | 1 - 2 | 50 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017000108.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
