Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017000107.1 | XP_016855596.1 | 6 - 7 | 576 | 66 | |
XM_017000108.1 | XP_016855597.1 | 8 - 9 | 958 | 66 | ||
GenBank mRNA | AK023586.1 | - | 3 - 4 | 209 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017000107.1, XM_017000108.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
