Assay Details
Gene Symbol: STAMBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017016454.1 | XP_016871943.1 | 2 - 3 | 288 | 68 | |
XM_017016455.1 | XP_016871944.1 | 2 - 3 | 965 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017016454.1, XM_017016455.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
