Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005245198.3 | XP_005245255.1 | 2 - 3 | 331 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005245198.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |

SERPINC1
Genomic Map