Assay Details
Gene Symbol: HHIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017002350.1 | XP_016857839.1 | 5 - 6 | 1542 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000494899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000494899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | ENST00000494899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000343410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000468172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000473144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000494899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000494899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | ENST00000494899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017435.hg.1 | NM_024746.3 | Refseq |
