Assay Details
| Gene Symbol: NVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002533.3 | NP_002524.2 | 5 - 6 | 409 | 128 | |
| NM_206840.2 | NP_996671.1 | 4 - 5 | 335 | 128 | ||
| XM_011544196.1 | XP_011542498.1 | 5 - 6 | 392 | 128 | ||
| XM_011544198.1 | XP_011542500.1 | 4 - 5 | 347 | 128 | ||
| XM_017001378.1 | XP_016856867.1 | 6 - 7 | 501 | 128 | ||
| XM_017001379.1 | XP_016856868.1 | 6 - 7 | 501 | 128 | ||
| XM_017001380.1 | XP_016856869.1 | 6 - 7 | 501 | 128 | ||
| XM_017001381.1 | XP_016856870.1 | 6 - 7 | 498 | 128 | ||
| XM_017001382.1 | XP_016856871.1 | 5 - 6 | 431 | 128 | ||
| XM_017001383.1 | XP_016856872.1 | 5 - 6 | 431 | 128 | ||
| XM_017001384.1 | XP_016856873.1 | 6 - 7 | 501 | 128 | ||
| XM_017001385.1 | XP_016856874.1 | 6 - 7 | 501 | 128 | ||
| GenBank mRNA | AK297070.1 | - | 5 - 6 | 368 | 128 | |
| AK298826.1 | - | 4 - 5 | 287 | 128 | ||
| AW997091.1 | - | 2 - 3 | 154 | 128 | ||
| BC005897.1 | - | 5 - 6 | 368 | 128 | ||
| BP284587.1 | - | 5 - 6 | 409 | 128 | ||
| U68140.1 | - | 5 - 6 | 403 | 128 | ||
| U78772.1 | - | 4 - 5 | 316 | 128 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK298826.1, NM_206840.2, U78772.1, XM_011544198.1 transcripts.
Set Membership: |
|

NVL
Genomic Map