Assay Details
| Gene Symbol: PPP1R15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032833.4 | NP_116222.4 | 2 | 4126 | 122 | |
| GenBank mRNA | AK023680.1 | - | 1 | 1010 | 122 | |
| AL833746.1 | - | 2 | 2802 | 120 | ||
| BC065280.1 | - | 2 | 4101 | 6172811 | 122 | |
More Information
Set Membership: |
|

PPP1R15B
Genomic Map