Assay Details
Gene Symbol: MINOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204083.1 | NP_001191012.1 | 4 - 5 | 532 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000479642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000479642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000479642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000322753.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000462646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000467029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000479642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000479642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000479642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000481464.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000485362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000486890.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000498642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | ENST00000617872.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001032363.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204082.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NM_001204083.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018185.hg.1 | NR_033758.2 | Refseq |
