Assay Details
| Gene Symbol: C1orf132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135298.1 | 2 - 3 | 674 | 119 | ||
| NR_135299.1 | 3 - 4 | 859 | 119 | |||
| GenBank mRNA | EU154351.1 | - | 2 - 3 | 674 | 119 | |
| EU154352.1 | - | 3 - 4 | 859 | 119 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000385055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000385055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000385231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000385231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | NR_029518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017137.hg.1 | NR_029832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000385055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000385055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000385231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000385231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000415882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000487977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | ENST00000608023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | NR_029518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017137.hg.1 | NR_029832.1 | Refseq |

C1orf132
Genomic Map