Assay Details
| Gene Symbol: LOC728323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130699.1 | 4 - 5 | 510 | 113 | ||
| NR_130700.1 | 3 - 4 | 407 | 113 | |||
| NR_130701.1 | 5 - 6 | 560 | 113 | |||
| GenBank mRNA | AK298283.1 | - | 4 - 5 | 450 | 113 | |
| AK307199.1 | - | 5 - 6 | 500 | 113 | ||
| AK309231.1 | - | 3 - 4 | 356 | 113 | ||
More Information
Set Membership: |
|

LOC728323
Genomic Map