Assay Details
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110767.1 | 2 - 3 | 399 | 99 | ||
GenBank mRNA | AK098044.1 | - | 2 - 3 | 384 | 99 | |
AL542628.3 | - | 2 - 3 | 386 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000411179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
