Assay Details
Gene Symbol: PDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK296288.1 | - | 1 - 2 | 182 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK296288.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000449300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000449300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000449300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000458171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000458171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000376215.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000449300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000449300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000449300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000458171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000458171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000470978.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000473224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | ENST00000491711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007081.hg.1 | NM_014317.4 | Refseq |
