Assay Details
| Gene Symbol: PLCG1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109889.1 | 2 - 3 | 397 | 95 | ||
| GenBank mRNA | BC035080.1 | - | 2 - 3 | 402 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLCG1-AS1| Gene Symbol: PLCG1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109889.1 | 2 - 3 | 397 | 95 | ||
| GenBank mRNA | BC035080.1 | - | 2 - 3 | 402 | 95 | |
Set Membership: |
|