| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000414189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
ENST00000436845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008141.hg.1 |
NR_110545.1 |
Refseq |