Assay Details
Gene Symbol: LOC101927973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126012.1 | 1 - 2 | 302 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000433303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000437194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | ENST00000453420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007841.hg.1 | NR_126012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000465684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000468072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000475886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | ENST00000497573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007667.hg.1 | NR_027104.1 | Refseq |
