Assay Details
Gene Symbol: CCKBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | Y13464.1 | - | 1 - 2 | 159 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
