Assay Details
Gene Symbol: CCKBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017018516.1 | XP_016874005.1 | 1 - 2 | 893 | 88 | |
GenBank mRNA | S76072.1 | - | 1 - 2 | 161 | 88 | |
Y13464.1 | - | 2 - 3 | 315 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the S76072.1, XM_017018516.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
gastric acid secretion
cell surface receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
digestion
sensory perception
feeding behavior
cell proliferation
positive regulation of cell proliferation
cholecystokinin signaling pathway
regulation of phosphatidylinositol 3-kinase activity
digestive tract development
gland development
phosphatidylinositol phospholipase C activity
cholecystokinin receptor activity
protein binding
gastrin receptor activity
type B gastrin/cholecystokinin receptor binding
1-phosphatidylinositol-3-kinase regulator activity
gastric acid secretion
cell surface receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
digestion
sensory perception
feeding behavior
cell proliferation
positive regulation of cell proliferation
cholecystokinin signaling pathway
regulation of phosphatidylinositol 3-kinase activity
digestive tract development
gland development
phosphatidylinositol phospholipase C activity
cholecystokinin receptor activity
protein binding
gastrin receptor activity
type B gastrin/cholecystokinin receptor binding
1-phosphatidylinositol-3-kinase regulator activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000334619.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000525462.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000531712.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | ENST00000532715.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006722.hg.1 | NM_176875.3 | Refseq |
