Assay Details
Gene Symbol: MPST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011530196.1 | XP_011528498.1 | 1 - 2 | 940 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
liver development
cyanate catabolic process
response to toxic substance
transsulfuration
spinal cord development
hydrogen sulfide biosynthetic process
thiosulfate sulfurtransferase activity
3-mercaptopyruvate sulfurtransferase activity
identical protein binding
kidney development
liver development
cyanate catabolic process
response to toxic substance
transsulfuration
spinal cord development
hydrogen sulfide biosynthetic process
thiosulfate sulfurtransferase activity
3-mercaptopyruvate sulfurtransferase activity
identical protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000341116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000341116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000341116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000341116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000341116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000397225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000397225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000397225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000397225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000397225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000401419.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000401419.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000401419.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000401419.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000401419.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404802.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404802.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404802.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404802.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000404802.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000429360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000429360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000429360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000429360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000485587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000628507.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | ENST00000628507.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001013436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001013436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001013436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001013436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001013436.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001130517.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001130517.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001130517.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001130517.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001130517.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_001130517.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_021126.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_021126.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_021126.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007277.hg.1 | NM_021126.5 | Refseq |
