Assay Details
| Gene Symbol: KDM4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033827.1 | 2 - 3 | 385 | 114 | ||
| GenBank mRNA | BC094867.1 | - | 2 - 3 | 385 | 114 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000418149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000418149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000418149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000398804.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000418149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000418149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000418149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000434346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000439057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | ENST00000453015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013935.hg.1 | NR_033827.1 | Refseq |

KDM4A-AS1
Genomic Map