Assay Details
| Gene Symbol: LSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021177.4 | NP_067000.1 | 3 - 4 | 330 | 165 | |
| GenBank mRNA | AF182288.1 | - | 3 - 4 | 149 | 165 | |
| AF196468.1 | - | 3 - 4 | 295 | 165 | ||
| AJ245416.1 | - | 3 - 4 | 321 | 165 | ||
| BC009192.2 | - | 3 - 4 | 312 | 3842537 | 165 | |
| BP203102.1 | - | 3 - 4 | 330 | 165 | ||
| BU189898.1 | - | 3 - 4 | 321 | 165 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF182288.1, AF196468.1, AJ245416.1, BC009192.2, NM_021177.4 transcripts.
Set Membership: |
|

LSM2
Genomic Map