Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006868.hg.1 |
ENST00000555922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
ENST00000368713.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015867.hg.1 |
NM_002960.1 |
Refseq |