Assay Details
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX099695.1 | - | 2 - 3 | 233 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000425537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000425537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000425537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000458277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000458277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000458277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NR_033264.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NR_033264.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007734.hg.1 | NR_033264.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000425537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000425537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000425537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000458277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000458277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000458277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000519684.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | ENST00000522796.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NM_145205.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NR_033264.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NR_033264.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007734.hg.1 | NR_033264.1 | Refseq |
