Assay Details
Gene Symbol: LINC00690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103843.1 | 2 | 255 | 75 | ||
GenBank mRNA | BC034913.1 | - | 2 | 258 | 75 | |
BX100558.1 | - | 2 | 255 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
