| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
ENST00000340087.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
ENST00000340087.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
ENST00000340087.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
ENST00000340087.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
ENST00000425699.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009057.hg.1 |
NM_199461.3 |
Refseq |