| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000611121.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
ENST00000612555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000500765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000511543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000611121.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
ENST00000612555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008110.hg.1 |
NR_002806.2 |
Refseq |