Assay Details
| Gene Symbol: ANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005262945.2 | XP_005263002.1 | 46 | 12176 | 120 | |
| XM_017008067.1 | XP_016863556.1 | 50 | 13227 | 120 | ||
| XM_017008068.1 | XP_016863557.1 | 51 | 13468 | 120 | ||
| XM_017008069.1 | XP_016863558.1 | 49 | 13202 | 120 | ||
| XM_017008070.1 | XP_016863559.1 | 50 | 13413 | 120 | ||
| XM_017008071.1 | XP_016863560.1 | 51 | 12396 | 120 | ||
| XM_017008072.1 | XP_016863561.1 | 50 | 12372 | 120 | ||
| XM_017008073.1 | XP_016863562.1 | 48 | 12368 | 120 | ||
| XM_017008074.1 | XP_016863563.1 | 50 | 12342 | 120 | ||
| XM_017008075.1 | XP_016863564.1 | 49 | 12306 | 120 | ||
| XM_017008076.1 | XP_016863565.1 | 47 | 12275 | 120 | ||
| XM_017008077.1 | XP_016863566.1 | 47 | 12183 | 120 | ||
| XM_017008078.1 | XP_016863567.1 | 46 | 6020 | 120 | ||
| XM_017008079.1 | XP_016863568.1 | 45 | 5843 | 120 | ||
| XM_017008080.1 | XP_016863569.1 | 46 | 5871 | 120 | ||
| GenBank mRNA | DQ925692.1 | - | 2 | 126 | 120 | |
| DQ925693.1 | - | 3 | 309 | 120 | ||
More Information
Set Membership: |
|

ANK2
Genomic Map