Assay Details
| Gene Symbol: LINC01134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130173.1 | - | 2 | 2216 | 99 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000413332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000439488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | ENST00000442673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006629.hg.1 | NR_024455.1 | Refseq |

LINC01134
Genomic Map