Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008916.hg.1 |
ENST00000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006896.hg.1 |
ENSTR0000437244.1 |
Ensembl |