Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
ENST00000378970.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009321.hg.1 |
NM_000475.4 |
Refseq |