Assay Details
| Gene Symbol: TSC22D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243799.1 | NP_001230728.1 | 1 | 1324 | 80 | |
| NM_183422.3 | NP_904358.2 | 1 | 1324 | 80 | ||
| XM_005266583.4 | XP_005266640.1 | 1 | 1187 | 80 | ||
| XM_017020810.1 | XP_016876299.1 | 1 | 1187 | 80 | ||
| XM_017020811.1 | XP_016876300.1 | 1 | 1187 | 80 | ||
| GenBank mRNA | AB082525.1 | - | 1 | 1324 | 80 | |
| AK091854.1 | - | 1 | 1015 | 80 | ||
| BC136475.1 | - | 1 | 1094 | 9020464 | 80 | |
| BC136482.1 | - | 1 | 1094 | 9020471 | 80 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000437455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000437455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000437455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000493016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000493016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000493016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000617014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000617014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | ENST00000617014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000616360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000617014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000617014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | ENST00000617014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | NR_038381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007039.hg.1 | NR_038381.1 | Refseq |

TSC22D1
Genomic Map