Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000369153.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
ENST00000476344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009881.hg.1 |
NM_016074.4 |
Refseq |