Assay Details
| Gene Symbol: STK35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080836.3 | NP_543026.2 | 4 | 4466 | 99 | |
| XM_011529174.2 | XP_011527476.1 | 3 | 7563 | 99 | ||
| GenBank mRNA | AK024394.1 | - | 1 | 2684 | 99 | |
More Information
Set Membership: |
|

STK35
Genomic Map