Assay Details
Gene Symbol: TMED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017022040.1 | XP_016877529.1 | 3 | 6106 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000561560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000561560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
