Assay Details
| Gene Symbol: SLCO4A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024470.1 | 4 | 69 | |||
| GenBank mRNA | BC025345.1 | - | 4 | 4862156 | 69 | |
| BU741282.1 | - | 1 | 138 | 69 | ||
More Information
Set Membership: |
|

SLCO4A1-AS1
Genomic Map