Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000433161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
ENST00000459583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008044.hg.1 |
NR_003244.1 |
Refseq |