Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000373170.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000448559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
ENST00000451810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007969.hg.1 |
NR_024015.1 |
Refseq |