Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |