Assay Details
| Gene Symbol: GPR50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004224.3 | NP_004215.2 | 2 | 971 | 72 | |
| XM_011531216.2 | XP_011529518.1 | 2 | 200 | 72 | ||
| GenBank mRNA | AY114102.1 | - | 2 | 128 | 72 | |
| BC103696.1 | - | 2 | 881 | 40006713 | 72 | |
| BC105683.1 | - | 2 | 881 | 40006718 | 72 | |
| BC105684.1 | - | 2 | 881 | 40006715 | 72 | |
| EU432118.1 | - | 2 | 833 | 72 | ||
| U52219.1 | - | 2 | 902 | 72 | ||
More Information
Set Membership: |
|

GPR50
Genomic Map