Assay Details
| Gene Symbol: GRIK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027021.1 | 2 | 362 | 84 | ||
| GenBank mRNA | AY077697.1 | - | 1 | 132 | 84 | |
| BG187681.1 | - | 2 | 367 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY077697.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000309331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000412579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000413131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000423221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | ENST00000455392.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006869.hg.1 | NR_027021.1 | Refseq |

GRIK1-AS1
Genomic Map