| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000593292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000593292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000593292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000593645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000593645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000593292.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000593292.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000593292.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000593645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000593645.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011924.hg.1 |
ENST00000595893.1 |
Ensembl |