Assay Details
Gene Symbol: CCDC106 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013301.2 | NP_037433.2 | 4 | 957 | 63 | ||
XM_005258827.2 | XP_005258884.1 | 4 | 555 | 63 | |||
XM_005258828.2 | XP_005258885.1 | 4 | 576 | 63 | |||
GenBank mRNA | AF054984.1 | - | 4 | 510 | 63 | ||
AK097095.1 | - | 3 | 1127 | 63 | |||
AY927563.1 | - | 1 | 590 | 63 | |||
AY927628.1 | - | LIT3614 AY927628 |
2 | 463 | 63 | ||
BC000412.2 | - | 3 | 535 | 2961290 | 63 | ||
BC008956.2 | - | 3 | 535 | 2961290 | 63 | ||
CR450308.1 | - | 3 | 222 | 63 | |||
GU354321.1 | - | 4 | 312 | 63 | |||
GU354322.1 | - | 4 | 368 | 63 | |||
GU354323.1 | - | 4 | 556 | 63 | |||
U79303.1 | - | 4 | 510 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000308964.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000586864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000587213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000588740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000591578.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | ENST00000593069.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011805.hg.1 | NM_013301.2 | Refseq |
