Assay Details
Gene Symbol: PREB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013388.4 | NP_037520.1 | 1 | 160 | 104 | |
XM_006711914.2 | XP_006711977.1 | 1 | 141 | 104 | ||
XM_011532471.2 | XP_011530773.1 | 1 | 141 | 104 | ||
XM_011532472.2 | XP_011530774.1 | 1 | 141 | 104 | ||
GenBank mRNA | AK001976.1 | - | 1 | 115 | 104 | |
AK023064.1 | - | 1 | 117 | 104 | ||
BC002765.2 | - | 1 | 91 | 3631721 | 104 | |
BC012890.1 | - | 1 | 101 | 4158577 | 104 | |
BC016472.1 | - | 1 | 91 | 4178911 | 104 | |
BC016906.1 | - | 1 | 101 | 4427483 | 104 | |
BC041032.1 | - | 1 | 154 | 4638313 | 104 | |
BG715668.1 | - | 1 | 166 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK023064.1, BC002765.2, BC012890.1, BC016472.1, BC016906.1, BC041032.1, NM_013388.4, XM_006711914.2, XM_011532471.2, XM_011532472.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
IRE1-mediated unfolded protein response
positive regulation of GTPase activity
COPII vesicle coating
DNA binding
guanyl-nucleotide exchange factor activity
Sar guanyl-nucleotide exchange factor activity
protein binding
transcription, DNA-templated
regulation of transcription, DNA-templated
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
IRE1-mediated unfolded protein response
positive regulation of GTPase activity
COPII vesicle coating
DNA binding
guanyl-nucleotide exchange factor activity
Sar guanyl-nucleotide exchange factor activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000416802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000416802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000416802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000260643.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000406567.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000416802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000416802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000416802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000430533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000441451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000444452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000456259.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000468045.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | ENST00000474802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012062.hg.1 | NM_013388.5 | Refseq |
