Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005246212.3 | XP_005246269.1 | 1 | 115 | 95 | |
| XM_011510459.2 | XP_011508761.1 | 1 | 243 | 95 | ||
| GenBank mRNA | AK302425.1 | - | 1 | 82 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302425.1, XM_005246212.3, XM_011510459.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000343105.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000409150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000414176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000421038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000444545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | ENST00000492292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015231.hg.1 | NM_001142645.1 | Refseq |

NEMP2
Genomic Map