Assay Details
| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303264.1 | NP_001290193.1 | 1 | 2854 | 103 | |
| NM_014779.3 | NP_055594.1 | 1 | 2854 | 103 | ||
| NR_130136.1 | 1 | 2854 | 103 | |||
| XM_011513337.2 | XP_011511639.1 | 1 | 2921 | 103 | ||
| GenBank mRNA | AB014569.1 | - | 1 | 2854 | 103 | |
| AF201290.1 | - | 1 | 210 | 103 | ||
| AK297383.1 | - | 2 | 748 | 103 | ||
| AK297748.1 | - | 1 | 183 | 103 | ||
| BC044643.1 | - | 1 | 2188 | 5454441 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK297748.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000460316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000466814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000480589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000485421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | ENST00000492828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_001303264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NM_014779.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013888.hg.1 | NR_130136.1 | Refseq |

TSC22D2
Genomic Map