| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |