Assay Details
Gene Symbol: TADA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152293.2 | NP_689506.2 | 2 | 3453 | 121 | |
XM_011513595.1 | XP_011511897.1 | 2 | 4133 | 121 | ||
GenBank mRNA | AK126966.1 | - | 2 | 3221 | 121 | |
BX648428.1 | - | 1 | 3026 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
histone acetylation
positive regulation of histone acetylation
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
zinc ion binding
chromatin remodeling
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
histone acetylation
positive regulation of histone acetylation
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000506692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000506692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000510704.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000510704.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000510704.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000221975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000593863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598466.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000598742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000600467.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | ENST00000601492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008164.hg.1 | NM_001022.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000310074.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000506692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000506692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000510704.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000510704.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000510704.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000512388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | ENST00000515646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006712.hg.1 | NM_152293.2 | Refseq |
