Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017007673.1 | XP_016863162.1 | 1 | 342 | 66 | |
GenBank mRNA | BC019894.1 | - | 1 | 622 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017007673.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
