Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013424.hg.1 |
ENST00000518602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013424.hg.1 |
ENST00000518602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000503428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000503428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000503428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000508188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000508188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000503428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000503428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000503428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000508188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000508188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013275.hg.1 |
ENST00000514793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013424.hg.1 |
ENST00000518602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013424.hg.1 |
ENST00000518602.1 |
Ensembl |