Assay Details
Gene Symbol: LOC553103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX538252.1 | - | 2 | 622 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
